High resolution colony counter, Scan® 1200
Leverandør: INTERSCIENCE
437000
INSC437000EA 0 NOK
INSC437000
High resolution colony counter, Scan® 1200
Kolonitellere
High resolution colony counter that can be linked to a PC via a FireWire connection.
  • Minimum size of colonies is 0,05 mm
  • Detects up to seven colours on the same Petri dish
  • Measurement of the inhibition zone
Can count colonies in less than one second for a fast, accurate, and traceable reading of the result.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Skålstørrelse 55 – 90 mm Petri dishes, Spiral® plating, Petrifi lm™, RIDA™ Count/Sanita-kun™, Compact Dry™ and filtration membrane
Elektronisk 240 V/50 – 60 Hz
Forstørrelse 28×
Kamera High definition CCD colour camera
Oppløsning 1280×960 pixels
B×D×H 280×270×380 mm
Vekt 9,4 kg