Pipetting robot, ASSIST PLUS
Leverandør: BDP International Inc.
4505
INTL4505EA 0 NOK
INTL4505
Pipetting robot, ASSIST PLUS
Pipetter Pipetter, repeterene stepper
INTEGRA has developed the ASSIST PLUS pipetting robot to streamline routine pipetting tasks. Using INTEGRA electronic multichannel pipettes, the system automates pipetting tasks, eliminates physical strain and ensures superior reproducibility and error free pipetting.
  • Automates any VOYAGER or VIAFLO multichannel electronic pipette
  • Compatible with all INTEGRA tube racks, reagent reservoirs and microplates (12-384 well)
  • Protocols are created either on the pipette or in VIALAB (free for download)
  • Easy and fast programming with the VIALAB pipetting software