Cleanroom Masks, Exa+ 250
Leverandør: ISOFIELD IRELAND
60411
ISOF60411EA 17925 NOK
ISOF60411
Cleanroom Masks, Exa+ 250
Masker Munnbind, renrom
Validated sterile cleanroom face mask with earloops and clip.
  • Large pouch-style breathing chamber
  • Extended lower breathing chamber provides excellent coverage at neck
  • Highly efficient bacterial and particle filtration
  • Gamma irradiation, min. 25 kGy
  • Particle shedding  (Helmke drum) Cat. 1
  • Sterility Assurance Level (SAL) 10⁻⁶
  • Cleanroom class: 10/ISO 4; EU GMP (A/B)
Sterile cleanroom face masks feature an ultra-wide 250 mm length with malleable nose strip.
Forpakning:
Masks are packaged 1 piece per sealed inner PE bag, 50 inner bags per sealed