VWR® UltraPure, Buffer, concentrate (phosphate buffered saline)
Leverandør: VWR Chemicals
J373-4LEA 5025 NOK
J373-4L
VWR® UltraPure, Buffer, concentrate (phosphate buffered saline)
Buffere Buffere, elektroforese Buffere, vask
  • pH at 25 °C (10% in water): 7,3 - 7,5
  • Conductivity (10% in water): 14000 - 17000 mhos
  • No DNases, RNases or proteases detected, sterile