VWR®, Pepstatin A
Leverandør: VWR Chemicals
J583-25MGEA 5625 NOK
J583-25MG
VWR®, Pepstatin A
Proteiner
An aspartic acid protease inhibitor of pepsin, renin, and cathepsin D. Also an inhibitor of the HIV protease. Not water soluble.