NP-40 lysis buffer with glycerol (2X)

Leverandør: Thermo Fisher Scientific

J60143.AK
J60143.AKEA 1125 NOK
J60143.AK
NP-40 lysis buffer with glycerol (2X)
Buffere Buffere, ekstrasjons

Storage Temperature: 2 to 8°C
Physical Form: Viscous liquid
Appearance: Colourless

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR