Okadaic acid ≥98%

Leverandør: Thermo Fisher Scientific
Danger

J60155.XS J60155.XG J60155.IX
J60155.XSEA 6825 NOK
J60155.XS J60155.XG J60155.IX
Okadaic acid ≥98%
Okadaic acid
Formel: C₄₄H₆₈O₁₃
Lagringstemperatur: Fryseskap
MDL nummer: MFCD00083455
CAS-nummer: 78111-17-8
FN: 3462
ADR: 6.1,I
Merck Indeks: 14,6819

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR