Azure A

Leverandør: Thermo Fisher Scientific
Warning

Synonymer: Azurol a, N,N-Dimethylthionine, Azure A chloride, azurol I, 7 - (dimethyl) - 3-imino-3h-dithizine hydrochloride, 3-amino-7-dimethylaminophenothiazine-5-onium chloride, 3-Amino-7-(dimethylamino)phenothiazin-5-ium chloride, 3-Amino-7-(dimethylamino)phenazathionium Chloride, Methylene Azur A

J61346.14 J61346.22
J61346.14EA 515 NOK
J61346.14 J61346.22
Azure A
Azure A

CAS No.: 531-53-3 Molecular Formula: C14H14ClN3S Formula Weight: 291.80 Physical Form: Powder Appearance: Dark green Melting Point: 290° dec. Beilstein Reference: 3922287 Merck Reference: 14,927 MDL No.: MFCD00012112


For staining cell granules and tissues

Formel: C₁₄H₁₄ClN₃S
MW: 291.797 g/mol
Smeltepunkt: 290 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00012112
CAS-nummer: 531-53-3
EINECS: 208-510-7

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR