Dicloxacillin sodium salt monohydrate

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: 3-(2,6-Dichlorophenyl)-5-methyl-4-isoxazolyl penicillin sodium salt monohydrate, Sodium (2S,5R,6R)-6-(3-(2,6-dichlorophenyl)-5-methylisoxazole-4-carboxamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate hydrate, Dicloxacillin Sodium hydrate

J61581.03 J61581.06
J61581.03EA 1025 NOK
J61581.03 J61581.06
Dicloxacillin sodium salt monohydrate
Dicloxacillin sodium salt monohydrate

CAS No.: 13412-64-1 Molecular Formula: C19H16Cl2N3NaO5S Formula Weight: 492.33 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Melting Point: 222-225° Solubility: Soluble in water MDL No.: MFCD00056865


Applications: An antibiotic used in the treatment of staphyloccoci bacteria that are resistant to penicillin G

Formel: C₁₉H₁₆Cl₂N₃NaO₅S·1H₂O
Smeltepunkt: 222…225 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00056865
CAS-nummer: 13412-64-1
EINECS: 603-794-2
Merck Indeks: 14,3084

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR