EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M vandig løsning, autoclaved pH 8.0

Leverandør: Thermo Fisher Scientific

J61721.ADEA 1450 NOK
J61721.AD
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) 0.5 M vandig løsning, autoclaved pH 8.0
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)

CAS No.: 67-42-5 Molecular Formula: C14H24N2O10 Formula Weight: 380.35 Physical Form: Liquid MDL No.: MFCD00004291

Formel: C₁₄H₂₄N₂O₁₀
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00004291
CAS-nummer: 67-42-5

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR