Lysis buffer, 0,5% Tween 20, pH 8,0, for isolation of His-tag modified proteins

Leverandør: Thermo Fisher Scientific

J61792.AP
J61792.APEA 855 NOK
J61792.AP
Lysis buffer, 0,5% Tween 20, pH 8,0, for isolation of His-tag modified proteins
Buffere Buffere, ekstrasjons

Physical Form: Liquid
MDL No.: MFCD00677267
Applications: For isolation of His-tag proteins

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR