Spectinomycin dihydrochloride pentahydrate cell culture reagent

Leverandør: Thermo Fisher Scientific

J61820.06 J61820.14
J61820.06EA 1625 NOK
J61820.06 J61820.14
Spectinomycin dihydrochloride pentahydrate cell culture reagent
Spectinomycin dihydrochloride pentahydrate

An aminocyclitol antibiotic that binds to the 30S ribosomal subunit of bacteria and interrupts protein synthesis


White powder.


Applications: An aminocyclitol antibiotic that binds to the 30S ribosomal subunit of bacteria and interrupts protein synthesis


Soluble in water

Formel: C₁₄H₂₄N₂O₇·2HCl·5H₂O
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00150886
CAS-nummer: 22189-32-8
EINECS: 244-554-3

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR