Carbenicillin disodium salt

Leverandør: Thermo Fisher Scientific
Danger

J61949.03 J61949.06 J61949.14
J61949.03EA 1325 NOK
J61949.03 J61949.06 J61949.14
Carbenicillin disodium salt
Carbenicillin disodium salt

CAS No.: 4800-94-6 Molecular Formula: C17H16N2Na2O6S Formula Weight: 422.36 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Beilstein Reference: 5722128 Merck Reference: 14,1792 MDL No.: MFCD00077683


Water-soluble antibiotic effective against Gram-negative bacteria

Formel: C₁₇H₁₆N₂O₆S·2Na
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00077683
CAS-nummer: 4800-94-6
EINECS: 225-360-8
Merck Indeks: 14,1792

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR