Sulindac Sulphide

Leverandør: Thermo Scientific
Danger

J62104.MCEA 4650 NOK
J62104.MC
Sulindac Sulphide
Sulindac Sulphide

CAS No.: 32004-67-4 Molecular Formula: C20H17FO2S Formula Weight: 340.41 Physical Form: Powder Appearance: Yellow Melting Point: 189-191° Solubility: Soluble in DMSO MDL No.: MFCD00869764


Applications: Active metabolite of sulindac; inhibits cyclooxygenase-1

Formel: C₂₀H₁₇FO₂S
MW: 340,4 g/mol
Smeltepunkt: 189…191 °C
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00869764
CAS-nummer: 32004-67-4
EINECS: 250-892-2

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR