Penicillin V potassium salt

Leverandør: Thermo Fisher Scientific
Danger

J62442.09 J62442.14 J62442.22
J62442.09EA 860 NOK
J62442.09 J62442.14 J62442.22
Penicillin V potassium salt
Penicillin V potassium salt

Inhibits bacterial cell wall synthesis.


White to off-white powder.


Applications: Inhibits bacterial cell wall synthesis

Formel: C₁₆H₁₇KN₂O₅S
Smeltepunkt: 197…202 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00051771
CAS-nummer: 132-98-9
EINECS: 205-086-5
Merck Indeks: 14,7100

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR