(S)-(+)-Camptothecin ≥90%

Leverandør: Thermo Fisher Scientific
Danger

J62523.MD J62523.03 J62523.06
J62523.MDEA 1375 NOK
J62523.MD J62523.03 J62523.06
(S)-(+)-Camptothecin ≥90%
(S)-(+)-Camptothecin

Cytotoxic antitumor agent that inhibits topoisomerase I.


Formula Weight: 348.35 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: Yellow Melting Point: 260° dec. Beilstein Reference: 631069 UN No.: UN1544 MDL No.: MFCD00081076


Applications: Cytotoxic antitumor agent that inhibits topoisomerase I

Formel: C₂₀H₁₆N₂O₄
Smeltepunkt: 260 °C (dec.)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00081076
CAS-nummer: 7689-03-4
FN: 1544
ADR: 6.1,III
Merck Indeks: 14,1735

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR