Erythrosin B, powder, spirit soluble

Leverandør: Thermo Fisher Scientific

Synonymer: C.I. 45430:2

J62619.06 J62619.14 J62619.22
J62619.06EA 480 NOK
J62619.06 J62619.14 J62619.22
Erythrosin B, powder, spirit soluble
Erythrosin B
Formel: C₂₀H₈I₄O₅
MW: 835.89 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00005044
CAS-nummer: 15905-32-5
EINECS: 240-046-0

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR