MES lysis buffer with Triton® X-100

Leverandør: Thermo Fisher Scientific

J62693.AK
J62693.AKEA 755 NOK
J62693.AK
MES lysis buffer with Triton® X-100
Buffere Buffere, ekstrasjons

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless

Bestill nå

SPESIFIKASJONER

Environmentally Preferable
utseende Colourless
Fyskisk form Liquid
Lagringstemp 2°C to 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR