Folic acid Cell Culture

Leverandør: Thermo Fisher Scientific

Synonymer: Vitamin M, (S)-2-(4-(((2-Amino-4-oxo-3,4-dihydropteridin-6-yl)methyl)amino)benzamido)pentanedioic acid, Pteroyl-L-glutamic acid, PteGlu, Vitamin B9

J62937.06 J62937.14 J62937.22
J62937.06EA 540 NOK
J62937.06 J62937.14 J62937.22
Folic acid Cell Culture
Folic acid

CAS No.: 59-30-3 Molecular Formula: C19H19N7O6 Formula Weight: 441.41 Physical Form: Powder Appearance: Yellow to orange Beilstein Reference: 100781 Merck Reference: 14,4221 MDL No.: MFCD00079305


Vitamin B9. A B-complex vitamin necessary for the body to produce red blood cells

Formel: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00079305
CAS-nummer: 59-30-3
EINECS: 200-419-0
Merck Indeks: 14,4221

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR