2'-Deoxycytidine

Leverandør: Thermo Fisher Scientific

J63062.03 J63062.06 J63062.14
J63062.03EA 510 NOK
J63062.03 J63062.06 J63062.14
2'-Deoxycytidine
2'-Deoxycytidine

CAS No.: 951-77-9 Molecular Formula: C9H13N3O4 Formula Weight: 227.22 Storage Temperature: 2°C to 8°C Physical Form: Crystalline powder Appearance: White Melting Point: 209-211° Beilstein Reference: 87567 MDL No.: MFCD00006547

Formel: C₉H₁₃N₃O₄
Smeltepunkt: 209…211 °C
MDL nummer: MFCD00006547
CAS-nummer: 951-77-9
EINECS: 213-454-1

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR