6-Cyano-7-nitroquinoxaline-2,3-dione 98%

Leverandør: Thermo Fisher Scientific
Warning

J63472.MA J63472.MB J63472.MF
J63472.MAEA 2250 NOK
J63472.MA J63472.MB J63472.MF
6-Cyano-7-nitroquinoxaline-2,3-dione 98%
6-Cyano-7-nitroquinoxaline-2,3-dione

Purity: 99+% CAS No.: 115066-14-3 Molecular Formula: C9H4N4O4 Formula Weight: 232.16 Storage Temperature: 2°C to 8°C Physical Form: Solid Melting Point: 300° Solubility: Soluble in DMSO to 100mM Beilstein Reference: 5821072 MDL No.: MFCD00069232


Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist

Formel: C₉H₄N₄O₄
Smeltepunkt: >300 °C
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00069232
CAS-nummer: 115066-14-3

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR