Kaliumklorid 1 M vandig løsning

Leverandør: Thermo Fisher Scientific

J63739.AE J63739.AK
J63739.AEEA 1050 NOK
J63739.AE J63739.AK
Kaliumklorid 1 M vandig løsning
Kaliumklorid

CAS No.: 7447-40-7 Molecular Formula: ClK Formula Weight: 74.55 Storage Temperature: 2°C to 8°C Physical Form: Liquid

Formel: KCl
Tetthet: 1,13 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00011360
CAS-nummer: 7447-40-7
EINECS: 231-211-8

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR