Penicillin G kaliumsalt

Leverandør: Thermo Fisher Scientific
Danger

J63901.14 J63901.22
J63901.14EA 760 NOK
J63901.14 J63901.22
Penicillin G kaliumsalt
Penicillin G kaliumsalt

CAS No.: 113-98-4 Molecular Formula: C16H17KN2O4S Formula Weight: 372.48 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White Beilstein Reference: 3832841 Merck Reference: 14,7094 MDL No.: MFCD00036193


Inhibits bacterial cell wall synthesis

Formel: C₁₆H₁₇KN₂O₄S
Smeltepunkt: 214…217 °C
Tetthet: 1,359 g/cm³ (20 °C)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00036193
CAS-nummer: 113-98-4
EINECS: 204-038-0
Merck Indeks: 14,7094

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR