(Hydroxy-2-naphthylmethyl)phosphonic acid 98%

Leverandør: Thermo Scientific
Warning

J63913.MB
J63913.MBEA 8250 NOK
J63913.MB
(Hydroxy-2-naphthylmethyl)phosphonic acid 98%
(Hydroxy-2-naphthylmethyl)phosphonic acid

Purity: 98% CAS No.: 120943-99-9 Molecular Formula: C11H11O4P Formula Weight: 238.18 Storage Temperature: -30°C to -10°C Physical Form: Solid Appearance: White MDL No.: MFCD00674174


Applications: A selective insulin receptor tyrosine kinase inhibitor

Formel: C₁₁H₁₁O₄P
MW: 238,18 g/mol
MDL nummer: MFCD00674174
CAS-nummer: 120943-99-9

Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR