2'-Deoxycytidine-5'-monophosphate 99%

Supplier: Thermo Fisher Scientific

J65244.03 J65244.06
J65244.03EA 1875 NOK
J65244.03 J65244.06
2'-Deoxycytidine-5'-monophosphate 99%
2'-Deoxycytidine-5'-monophosphate

Purity: 99% CAS No.: 1032-65-1 Molecular Formula: C9H14N3O7P Formula Weight: 307.20 Storage Temperature: -30°C to -10°C Physical Form: Crystalline powder Appearance: White Beilstein Reference: 41062 MDL No.: MFCD00006546

Formula: C₉H₁₄N₃O₇P
Storage Temperature: Freezer
MDL Number: MFCD00006546
CAS Number: 1032-65-1
EINECS: 213-849-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR