Toluidinblått O >65% (dye content), certified

Leverandør: Thermo Fisher Scientific

Synonymer: TBO, 3-Amino-7-(dimethylamino)-2-methyl-5-phenothiazinium chloride, 3-Amino-7-(dimethylamino)-2-methylphenothiazin-5-ium chloride, Tolonium chloride, Basic Blue 17, Blutene chloride

J66015.06 J66015.14
J66015.06EA 375 NOK
J66015.06 J66015.14
Toluidinblått O >65% (dye content), certified
Toluidinblått O

CAS No.: 92-31-9 Molecular Formula: C15H16ClN3S Formula Weight: 305.83 Physical Form: Powder Appearance: Dark green Odour: Odourless Solubility: Soluble in water at 1mg/ml MDL No.: MFCD00011934

Formel: C₁₅H₁₆ClN₃S
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011934
CAS-nummer: 92-31-9
EINECS: 202-146-2

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR