Spectinomycin sulphate 93%

Leverandør: Thermo Fisher Scientific
Danger

J66034.03 J66034.06
J66034.03EA 610 NOK
J66034.03 J66034.06
Spectinomycin sulphate 93%
Spectinomycin sulphate

This spectinomycin sulfate antibiotic acts against Gram-negative bacteria.


  • White powder


Soluble in DMSO or water

Formel: C₁₄H₂₄N₂O₇·H₂SO₄
Kokepunkt: 583 °C
Flash punkt: >110 °C (230 °F)
Lagringstemperatur: Kjøleskap
MDL nummer: MFCD00270185
CAS-nummer: 23312-56-3
EINECS: 245-578-7

Bestill nå

SPESIFIKASJONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR