Piperacillin sodium salt 835-1007 µg/mg

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: Piperacillin sodium, Sodium (2S,5R,6R)-6-((R)-2-(4-ethyl-2,3-dioxopiperazine-1-carboxamido)-2-phenylacetamido)-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate

J66419.06 J66419.09
J66419.06EA 3275 NOK
J66419.06 J66419.09
Piperacillin sodium salt 835-1007 µg/mg
Piperacillin sodium salt

CAS No.: 59703-84-3 Molecular Formula: C23H26N5NaO7S Formula Weight: 539.54 Storage Temperature: 2°C to 8°C Physical Form: Powder Appearance: White to off-white Odour: Odourless Melting Point: 183-185° Boiling Point: 793° Solubility: Soluble in water or methanol Beilstein Reference: 5373920 MDL No.: MFCD00917471


Applications: A broad spectrum antibiotic targeting a wide range of susceptible gram positive and gram negative bacteria

Formel: C₂₃H₂₇N₅O₇S
MW: 539.54 g/mol
Kokepunkt: 793 °C
Smeltepunkt: 183…185 °C
MDL nummer: MFCD00056867
CAS-nummer: 59703-84-3
EINECS: 261-868-6

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR