Etidiumbromid 98% for molecular biology

Leverandør: Thermo Fisher Scientific
Danger

Synonymer: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide

J67270.14
J67270.14EA 10925 NOK
J67270.14
Etidiumbromid 98% for molecular biology
Etidiumbromid

Purity: 98% CAS No.: 1239-45-8 Molecular Formula: C21H20BrN3 Formula Weight: 394.32 Physical Form: Powder Appearance: Red to purple Odour: Weak Melting Point: ca 261° dec. Beilstein Reference: 3642536 UN No.: UN2811 MDL No.: MFCD00011724

Formel: C₂₁H₂₀BrN₃
MW: 394.31 g/mol
Kokepunkt: 72
Smeltepunkt: ∼ 261 °C (dec.)
Lagringstemperatur: Romtemperatur
MDL nummer: MFCD00011724
CAS-nummer: 1239-45-8
EINECS: 214-984-6
FN: 2811
ADR: 6.1 I
Merck Indeks: 13,04751

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR