VWR® LB, Broth, Liquid, for Biotechnology
Leverandør: VWR Chemicals
J833-1LEA 1450 NOK
J833-1L
VWR® LB, Broth, Liquid, for Biotechnology
Mikrobiologiske medier Media, preparerte for mikrobiologi
LB Broth, liquid is used for the propagation and maintenance of E. coli. Widely used for the preparation of plasmid DNA and recombinant proteins.
  • Ready to use liquid media
  • Clear, yellow to brown liquid
One L of media contains 10 g sodium chloride, 10 g tryptone/case in peptone, 5 g yeast extract, bacteriological.
No growth detected in bioburden (Milliflex) (EQC) test.
pH (25 °C): 6,65 to 7,15.