VWR®, Malt extract, bacteriological grade
Leverandør: VWR Chemicals
J873-500GEA 2750 NOK
J873-500G
VWR®, Malt extract, bacteriological grade
Mikrobiologiske medier Media, dehydrert for mikrobiologi
Used for the preparation of media for the detection of yeasts and moulds.