Multi-parameter meter (pH/conductivity), bench, 3540
Leverandør: Cole-Parmer
354001 79000-58
662-0331EA 20000 NOK
662-0331 JENW79000-58
Multi-parameter meter (pH/conductivity), bench, 3540
Elektrokjemiske målere pH-meter
Ideal for use in all laboratories where pH and conductivity analyses are required.
  • Simultaneous display of pH and conductivity
  • Two independent channels to prevent interference between probes
  • Additional modes for resistivity, salinity, TDS and mV
  • Data logger with auto save/print options
  • Storage of up to 500 readings (250 for each mode)
Bestillingsinformasjon: Supplied with glass combination pH electrode 662-0343, glass conductivity probe (K=1, 663-0005), electrode stand and holder, ATC probe 662-0338, BNC shorting plug, pH buffers and power supply.