Safety air outlet filter
Leverandør: VICI JOUR RESEARCH
JR-S-20004
JOURJR-S-20004EA 1500 NOK
JOURJR-S-20004
Safety air outlet filter
Filter Filter, inntaks-
This safety air outlet filter regulates the pressure during helium sparging.