Impact goggle, Martcare®
Leverandør: JSP
AGC010-301-300
JSPAAGC010-301-300EA 62 NOK
JSPAAGC010-301-300
Impact goggle, Martcare®
Vernebriller /Goggles
A cost-effective, compact, and lightweight goggle.
  • Compact and lightweight
  • Tough polycarbonate lens
  • Impact protection
  • Vented body
  • PVC frame
A compact and lightweight goggle with tough polycarbonate lens and PVC frame, offering high resistance to impact from flying particles.
Sertifiseringer: Conforms to EN 166.1.B.