Elasticated chinstrap for JSP helmets
Leverandør: JSP
AHV001-100-000
JSPAAHV001-100-000EA 535 NOK
JSPAAHV001-100-000
Elasticated chinstrap for JSP helmets
Hjelmer Sikkerhethjelm
Elasticated chinstrap releases automatically with forces between 150 and 250 Newtons.
  • Wide elastic chinstrap
  • Releases automatically with forces between 150 and 250 Newtons
  • Nylon and elastic webbing
  • HDPE clips
Fits all Evolution® safety helmets.
Sertifiseringer: EN 397, EN 812.