Cough Guard 2 (fits bump caps)
Leverandør: JSP
AHV970-001-100
JSPAAHV970-001-100EA 335 NOK
JSPAAHV970-001-100
Cough Guard 2 (fits bump caps)
Ansiktsskjermer
The Cough Guard can be fitted to a JSP bump cap to reduce the likelihood of exhaled bacteria/virus contacting the wearer's face. This device is not specifically certified and tested for COVID-19 protection.
  • Crystal clear APET 0,3 mm lens provides a layer of protection
  • The only JSP verified cough guard bump cap fitment
  • Quick and simple cleaning with soap and water
  • Prescription eyewear, face mask and ear defender compatible
Crystal clear APET 0,3 mm lens provides a layer of protection, helping to block aerosols and droplets. The device easily fits JSPMore Product Information