Sweatband, Chamlon™

Leverandør: JSP
AJA840-000-200
JSPAAJA840-000-200EA 415 NOK
JSPAAJA840-000-200
Sweatband, Chamlon™
Hjelmer Sikkerhethjelm
Replacement Chamlon™ sweatband.

  • For use with the EVO® range of safety helmets
  • pH neutral
  • Dermatologically tested

Replacement Chamlon sweatband for use with the EVO® range of safety helmets. Egyptian cotton core with porous PU coating for maximum sweat absorption.
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR