Industrial working at height safety helmet, EVOLite® Skyworker™
Leverandør: JSP
AJS260-000-100
JSPAAJS260-000-100EA 1600 NOK
JSPAAJS260-000-100
Industrial working at height safety helmet, EVOLite® Skyworker™
Hjelmer Sikkerhethjelm
The EVOLite® Skyworker™ helmet has a suspension system and shell structure that is designed for use in industrial, mountaineering, rescue, and leisure activities.
  • Industrial working at height safety helmet
  • Meets the EN 12492 mountaineering standard
  • Strong lightweight ABS shell
  • EPS liner for side, front, and rear impact protection
  • Fully adjustable 4-point chinstrap with quick release buckle
The EVOLite® Skyworker™ helmet conforms to the mandatory shock absorption and penetration requirements of EN 397 industrial standard. The helmet features a fullyMore Product Information