Dust industrial PAPR kit, Jetstream®
Leverandør: JSP
CBP030-001-300
JSPACBP030-001-300EA 23600 NOK
JSPACBP030-001-300
Dust industrial PAPR kit, Jetstream®
Respiratorer Vernemaskesystem
The Jetstream® Dust Industrial Kit consists of a belt-mounted rechargeable respirator with a large capacity disposable filter supplying clean air through a highly flexible reinforced hose to a grinding visor headtop.
  • Grinding visor headtop
  • No face fit required
  • Flexible hose
  • PSL dust filters
  • Polycarbonate visor
  • Multi-region plug adaptor
The Jetstream® Industrial kit contains: grinding visor headtop, flexible hose, PSL filters, flow meter, rechargeable battery, and multi-region plug adaptor.
Sertifiseringer: EN 12941; EN166.