Expander barrier, 3 m, Titan®
Leverandør: JSP
KAZ110-005-300
JSPAKAZ110-005-300EA 4300 NOK
JSPAKAZ110-005-300
Expander barrier, 3 m, Titan®
Trafikksikkerhet Trafikkjegle/-barriere
Expandable and robust barrier providing a width of over 3 metres when opened to the maximum.
  • Lightweight, compact, and flexible
  • Just 55cm wide when closed; extremely practical for transporting
  • Opens out to over 3 metres wide
When closed the barrier is just 550mm, which makes it extremely practical for transporting. Expanding action gives user length flexibility fixed-length barriers systems do not have. Overall weight of 10.5kg means it is light enough to handle with relative ease, but stable enough to hold itself upright in windy conditions, roughMore Product Information