CORIO™ CP Refrigerated/Heating Circulators
Leverandør: JULABO GmbH
9013701.33 9013701.N1
JULA9013701.33EA 55450 NOK
JULA9013701.33 JULA9013701.N1
CORIO™ CP Refrigerated/Heating Circulators
Termostatbad Termostatbad, åpent Termostatbad / termostatbad med kjøling
CORIO™ CP refrigerated/heating circulator combinations with stainless steel bath tanks for internal or external temperature applications from –50 to +200 °C.
  • Infinitely adjustable pressure pump
  • Easy to switch between internal and external circulation
  • Bright LED display
  • USB and RS232 ports
For internal and external temperature applications. High quality stainless steel bath tanks with bath lid and drain tap.
CORIO CP circulators feature a bright LED display button controls. A lever below the display enables simple adjustment between internal andMore Product Information