Notrax® 470 Marble Sof-Tyle™ Floor Mattings
Leverandør: JUSTRITE MANUFACTURING
470S2436GY
JUSU470S2436GYEA 1550 NOK
JUSU470S2436GY
Notrax® 470 Marble Sof-Tyle™ Floor Mattings
Gulvmatter Gulvmatter, støtdempende
Marble Sof-Tyle™ is an anti-fatigue floor mat provides comfort and ergonomic support and complements any décor with a stylish smooth marble-pattern design.
  • Heavy-duty vinyl marble-pattern top surface combined with a dense closed cell PVC foam base
  • Smooth attractive surface is easy to sweep clean
  • UniFusion™ bond guaranteed for the life of the mat
  • Redstop™ non-slip backing to reduce mat slippage
  • Beveled edges on all 4 sides
  • Certified by the NFSI (National Floor Safety Institute)
The durable vinyl top surface is easy to sweep clean or damp mop, and isMore Product Information