549 Safety Stance™ Anti-fatigue Mats, Heavy Duty
Leverandør: JUSTRITE MANUFACTURING
549S3124OB
JUSU549S3124OBEA 5200 NOK
JUSU549S3124OB
549 Safety Stance™ Anti-fatigue Mats, Heavy Duty
Gulvmatter Gulvmatter, støtdempende
High quality anti-fatigue matting for use in areas where there can be frequent oil spilling.
  • Made of 100% nitrile rubber compound
  • Designed to withstand industrial oils and harsh chemicals
  • Suited for multi-shift operations
  • Aggressive slip-resistant surface
  • Recommended use: Heavy duty wet/dry industrial environments
Drainage holes and raised studs provide aeration and allow fluids and debris to fall through, leaving a dry and tidy top surface. Ergonomic benefit derived from 22 mm thick resilient worker platform. Highly visible nitrile rubber orangeMore Product Information