VWR® Transfer Buffer Concentrate, CAPS 10X
Leverandør: VWR Chemicals
K872-500MLEA 9675 NOK
K872-500ML
VWR® Transfer Buffer Concentrate, CAPS 10X
Buffere Buffere, elektroforese Buffere, transfer
For transfer of protein to membranes for sequencing or blotting.