Trypan blue 4 g/l in aqueous solution cell culture reagent

Leverandør: VWR Chemicals
Danger

Synonymer: Tolidine-disazo-bis-8-amino-1-naphthol-3,6-disulphonic acid tetrasodium salt, CI 23850, 3,3′-Dimethyl-4,4′-bis(5-amino-4-hydroxy-2,7-disulphonaphthyl-3-azo)-[1,1′-biphenyl] tetrasodium salt, Benzamine blue, Trypan blue sodium salt, Direct Blue 14, Diphenyl Blue, AzidinBlue, Sodium 3,3'-((3,3'-dimethyl-[1,1'-biphenyl]-4,4'-diyl)bis(diazene-2,1-diyl))bis(5-amino-4-hydroxynaphthalene-2,7-disulphonate)

K940-100MLEA 1025 NOK
K940-100ML
Trypan blue 4 g/l in aqueous solution cell culture reagent
Trypan blue

A pre-mixed dye solution used in cell culture applications to determine cell viability. A researcher can remove a sample of cells from culture and combine in a 1:1 ratio with the Trypan Blue solution. Under the microscope, dead cells will appear a blue color and viable cells will appear clear and translucent. Using a hemacytometer (gridded microscope slide), a researcher can quantify the percentage of dead cells within a population. Stained cells are ready for counting within five minutes.

Formel: C₃₄H₂₄N₆Na₄O₁₄S₄
MDL nummer: MFCD00003969
CAS-nummer: 72-57-1
EINECS: 200-786-7

Bestill nå

Spesifikasjon Testresultater

Abs.@ Lambda Max (1:100, Water) 1 - 1.3
Lambda Max (1:100, Water) 585 - 595 nm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR