PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Leverandør: KEYSTONE CLEANROOM PRODUCTS
10TK12
KACC10TK12EA 58500 NOK
KACC10TK12
PureClean™ Tyvek® 1421B Autoclavable Muff Covers
Deksler
PureClean™ Autoclavable muff covers are made from 1421B DuPont™ Tyvek®, which is a new grade of uncoated, untreated, flexible Tyvek® material. It has been specifically designed for pharmaceutical use in steam sterilisation applications. PureClean™ Tyvek® material does not have a water soluble antistat agent applied to it during its manufacturing process.
  • Uncoated Tyvek® for maximum breathability during sterilisation
  • Eliminate the risk of residue deposits on equipment post sterilisation
  • Manufactured and packaged in a controlled cleanroom environment
  • Reduces
More Product Information
New Product