Sterile Cleanroom Face Masks with Tyvek® Side Extensions
Leverandør: KEYSTONE CLEANROOM PRODUCTS
1461STXLDSM25 1461STXLDSMA25
KACC1461STXLDSM25EA 25400 NOK
KACC1461STXLDSM25 KACC1461STXLDSMA25
Sterile Cleanroom Face Masks with Tyvek® Side Extensions
Masker Munnbind, renrom
Sterile cleanroom face masks feature Tyvek® side extensions to prevent skin exposure. Double elastic headband fastening makes it easier to put on compared to ties. Optional fog-free design prevents goggles from fogging and mask slippage.
  • Tyvek® side extensions provide wider coverage area and an improved fit, while also eliminating exposed skin
  • Optional adhesive nose strip prevents goggles from fogging
  • Innovative design prevents mask slippage, reducing contamination risks and significant material and labour costs associated with re-gowning
  • Prevents the