Tyvek Sleeve Cover, Sterile
Leverandør: KEYSTONE CLEANROOM PRODUCTS
1804TW28CFDPST
KACC1804TW28CFDPSTEA 39900 NOK
KACC1804TW28CFDPST
Tyvek Sleeve Cover, Sterile
Engangsplagg
Extra-long sterile Tyvek sleeves are packaged in multiple layers of Tyvek peel pouches and poly bags to gain access into the aseptic area without excessive spraying and wiping with sanitisation agents. The sleeves are individually packed and specially folded for easy aseptic donning.
  • Manufactured and packaged in an ISO certified cleanroom
  • Clean and lint free
  • Tear and puncture resistant
  • Provides an excellent barrier to particles and microorganisms
  • Heat-sealed seams eliminate particles and liquids from passing through the seams
  • Each sleeve has elastic at the