Gamma Irradiation Sterilisation Bags, Self-Seal
Leverandør: KEYSTONE CLEANROOM PRODUCTS
3312IRD 3315IRD 3320IRD 3322IRD 3330IRD 3334IRD 3335IRD
KACC3312IRDEA 4525 NOK
KACC3312IRD KACC3315IRD KACC3320IRD KACC3322IRD KACC3330IRD KACC3334IRD KACC3335IRD
Gamma Irradiation Sterilisation Bags, Self-Seal
Poser Steriliseringsposer
Gamma irradiation sterilisation bags can be used as the primary, secondary, or tertiary packaging of items that will go through gamma irradiation processing, or sterilisation.
  • Appropriate for packaging of items that will be exposed to gamma radiation dose ranges between 15 and 50 kGy
  • After irradiation processing the process indicator will turn from yellow to red
  • The bags will provide protection of the contents from environmental factors, which could compromise sterility
  • Made of a transparent, strong, 3 mil thick medical grade polyolefin material with a