Biohazard Bags, Autoclavable, Sterile, Double Pouched
Leverandør: KEYSTONE CLEANROOM PRODUCTS
3748CBHNPST10D
KACC3748CBHNPST10DEA 6850 NOK
KACC3748CBHNPST10D
Biohazard Bags, Autoclavable, Sterile, Double Pouched
Poser Avfallsposer
Keystone Cleanroom Products Pre-sterilised biohazard bags are useful to decontaminate waste or potentially hazardous materials that are exiting the aseptic production area through the autoclave. The use of sterile biohazard bags eliminates the introduction of contamination into the aseptic production area.
  • Ready to use, packed in easy to open Chevron-Peel DuPont™ Tyvek® pouches
  • Maximum sterilisation temperature: 135 °C (275 °F)
  • Each package is labeled with an expiration date and lot number for complete traceability
Eliminate a common source ofMore Product Information